siRNA Details
| Gene Symbol: TMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135820.1 | NP_001129292.1 | 11, 12 | 2612 | ||
| NM_013390.2 | NP_037522.1 | 12, 13 | 2801 | |||
| XM_005251869.4 | XP_005251926.1 | 12, 13 | 2434 | |||
| GenBank mRNA | AB037833.2 | - | 12, 13 | 2394 | ||
| AF137030.1 | - | 12, 13 | 2409 | |||
| AK075370.1 | - | 5, 6 | 529 | |||
| BC140773.1 | - | 11, 12 | 2114 | 9021641 | ||
| BC144510.1 | - | 12, 13 | 2269 | |||
| BC146780.1 | - | 12, 13 | 2394 | 9007233 | ||
| BC171782.1 | - | 11, 12 | 2115 | |||

TMEM2
Genomic Map