siRNA Details
| Gene Symbol: SMPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014332.2 | NP_055147.1 | 3 | 344 | ||
| NR_045617.1 | 3 | 344 | ||||
| GenBank mRNA | AF129505.1 | - | 3 | 296 | ||
| AJ250584.1 | - | 3 | 296 | |||
| AK312134.1 | - | 3 | 344 | |||
| BC005948.1 | - | 3 | 225 | 4246501 | ||
| BF126275.1 | - | 3 | 206 | |||

SMPX
Genomic Map