siRNA Details
| Gene Symbol: KIAA0907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014949.3 | NP_055764.2 | 13 | 1691 | ||
| GenBank mRNA | AB020714.1 | - | 13 | 1648 | ||
| AY112680.1 | - | 13 | 1622 | |||
| AY112681.1 | - | 13 | 1622 | |||
| BC062637.1 | - | 13 | 1646 | 5497581 | ||

KIAA0907
Genomic Map