siRNA Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181718.3 | NP_859069.2 | 3 | 1374 | ||
| XM_017023107.1 | XP_016878596.1 | 4 | 1355 | |||
| GenBank mRNA | BC009033.1 | - | 3 | 1101 | ||
| BC029136.1 | - | 3 | 810 | |||
| BC034275.3 | - | 3 | 847 | |||
| BC040933.2 | - | 3 | 886 | |||
| BC085011.1 | - | 3 | 1222 | |||

ASPHD1
Genomic Map