siRNA Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181718.3 | NP_859069.2 | 2 | 1180 | ||
| XM_017023107.1 | XP_016878596.1 | 3 | 1161 | |||
| GenBank mRNA | AF070642.1 | - | 2 | 521 | ||
| BC009033.1 | - | 2 | 907 | |||
| BC029136.1 | - | 2 | 616 | |||
| BC034275.3 | - | 2 | 653 | |||
| BC040933.2 | - | 2 | 692 | |||
| BC085011.1 | - | 2 | 1028 | |||
| BC126319.1 | - | 2 | 1148 | 8992035 | ||
| BC126321.1 | - | 2 | 1148 | 8992037 | ||
| BC144152.1 | - | 2 | 1151 | 9052672 | ||

ASPHD1
Genomic Map