siRNA Details
Gene Symbol: STEAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207342.2 | NP_997225.1 | 5 | 1125 | ||
GenBank mRNA | BC066301.1 | - | 5 | 1022 | 5274319 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: STEAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207342.2 | NP_997225.1 | 5 | 1125 | ||
GenBank mRNA | BC066301.1 | - | 5 | 1022 | 5274319 |