siRNA Details
| Gene Symbol: ASF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014034.2 | NP_054753.1 | 4 | 898 | ||
| GenBank mRNA | AB028628.1 | - | 4 | 724 | ||
| AF151856.1 | - | 6 | 1175 | |||
| AF161495.1 | - | 6 | 1175 | |||
| AF279306.1 | - | 4 | 710 | |||
| AK025738.1 | - | 4 | 755 | |||
| AL050261.1 | - | 4 | 753 | |||
| BC010878.1 | - | 4 | 737 | 3546858 | ||
| CR457347.1 | - | 4 | 561 | |||

ASF1A
Genomic Map