siRNA Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014077.3 | NP_054796.1 | 2, 3 | 257 | ||
| GenBank mRNA | AF151902.1 | - | 2, 3 | 216 | ||
| AK312058.1 | - | 2, 3 | 240 | |||
| AL050157.1 | - | 2, 3 | 235 | |||
| BC000639.1 | - | 2, 3 | 235 | 3345441 | ||
| BC004447.1 | - | 1, 2 | 87 | |||
| BC017286.1 | - | 2, 3 | 218 | 4892079 | ||
| BG749750.1 | - | 1, 2 | 384 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026455, AB041658, BC006058, AK008336, AK011922, and others.

FAM32A
Genomic Map