siRNA Details
| Gene Symbol: FAM127B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078172.1 | NP_001071640.1 | 1 | 858 | ||
| NM_001134321.1 | NP_001127793.1 | 2 | 310 | |||
| GenBank mRNA | AL117556.1 | - | 1 | 797 | ||
| BC000393.2 | - | 1 | 831 | 2821721 | ||
| BC008096.1 | - | 1 | 616 | |||
| BC019300.1 | - | 1 | 831 | 2821721 | ||
| BC071991.1 | - | 2 | 221 | |||
| CN367136.1 | - | 2 | 264 | |||
| CR457123.1 | - | 1 | 174 | |||
| CR542228.1 | - | 1 | 174 | |||

FAM127B
Genomic Map