siRNA Details
| Gene Symbol: OR7A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017506.1 | NP_059976.1 | 1 | 815 | ||
| XM_011527912.2 | XP_011526214.1 | 2 | 840 | |||
| XM_011527913.2 | XP_011526215.1 | 2 | 766 | |||
| XM_017026583.1 | XP_016882072.1 | 2 | 729 | |||
| GenBank mRNA | AK312475.1 | - | 2 | 681 | ||
| BC104809.1 | - | 1 | 650 | 8143812 | ||
| X64976.1 | - | 1 | 218 | |||
| X89677.1 | - | 1 | 218 | |||

OR7A5
Genomic Map