siRNA Details
| Gene Symbol: OR4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001912.2 | NP_001001912.2 | 1 | 578 | ||
| GenBank mRNA | AY726598.1 | - | 1 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4E2| Gene Symbol: OR4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001912.2 | NP_001001912.2 | 1 | 578 | ||
| GenBank mRNA | AY726598.1 | - | 1 | 526 | ||