siRNA Details
| Gene Symbol: RPH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143854.1 | NP_001137326.1 | 20 | 2251 | ||
| NM_014954.3 | NP_055769.2 | 19 | 2239 | |||
| XM_011538048.1 | XP_011536350.1 | 20 | 2055 | |||
| XM_017019034.1 | XP_016874523.1 | 12 | 1509 | |||
| GenBank mRNA | AB023202.1 | - | 20 | 2172 | ||
| AK295696.1 | - | 20 | 2205 | |||
| AK296296.1 | - | 18 | 1790 | |||
| AK316112.1 | - | 20 | 2086 | |||
| BC017259.2 | - | 19 | 2046 | 3510158 | ||
| BX647281.1 | - | 12 | 2845 | |||

RPH3A
Genomic Map