siRNA Details
| Gene Symbol: BRICD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182563.3 | NP_872369.2 | 3 | 477 | ||
| XM_005255263.4 | XP_005255320.1 | 3 | 2446 | |||
| GenBank mRNA | AY597811.1 | - | 3 | 242 | ||
| BC039154.2 | - | 3 | 419 | 4213860 | ||

BRICD5
Genomic Map