siRNA Details
| Gene Symbol: FAM19A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004440.1 | NP_001004440.1 | 5 | 582 | ||
| NM_182759.2 | NP_877436.1 | 5 | 514 | |||
| XM_005270766.3 | XP_005270823.1 | 6 | 801 | |||
| XM_006710582.3 | XP_006710645.1 | 6 | 869 | |||
| GenBank mRNA | AY325116.1 | - | 5 | 514 | ||
| AY325119.1 | - | 5 | 582 | |||
| BC112268.1 | - | 5 | 576 | 8327736 | ||
| BC117479.1 | - | 5 | 576 | 40126030 | ||
| BC143949.1 | - | 5 | 545 | |||
| BC144042.1 | - | 5 | 565 | |||
| BC144043.1 | - | 5 | 508 | 9052563 | ||

FAM19A3
Genomic Map