siRNA Details
| Gene Symbol: LOC284912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001801.1 | - | 2 | 412 | 3355513 | |
| BG471420.1 | - | 2 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284912| Gene Symbol: LOC284912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001801.1 | - | 2 | 412 | 3355513 | |
| BG471420.1 | - | 2 | 413 | |||