siRNA Details
| Gene Symbol: AAMDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316960.1 | NP_001303889.1 | 5 | 639 | ||
| NM_001316961.1 | NP_001303890.1 | 5 | 559 | |||
| NM_001316962.1 | NP_001303891.1 | 5 | 563 | |||
| NM_024684.3 | NP_078960.1 | 4 | 521 | |||
| XM_011544968.2 | XP_011543270.1 | 4 | 521 | |||
| XM_011544969.2 | XP_011543271.1 | 4 | 514 | |||
| GenBank mRNA | AK024688.1 | - | 4 | 480 | ||
| BC002752.2 | - | 4 | 473 | 3628061 | ||
| BM930331.1 | - | 5 | 508 | |||

AAMDC
Genomic Map