siRNA Details
| Gene Symbol: STRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083893.1 | NP_001077362.1 | 5 | 776 | ||
| NM_014574.3 | NP_055389.3 | 5 | 776 | |||
| XM_005267569.3 | XP_005267626.1 | 5 | 903 | |||
| XM_005267570.3 | XP_005267627.1 | 5 | 903 | |||
| GenBank mRNA | AF243424.1 | - | 4 | 205 | ||
| AK290229.1 | - | 5 | 754 | |||
| AK314041.1 | - | 5 | 742 | |||
| BC126221.1 | - | 5 | 694 | 8991937 | ||
| BC132673.1 | - | 5 | 694 | 40146695 | ||
| BC143933.1 | - | 5 | 694 | |||
| U17989.1 | - | 5 | 764 | |||

STRN3
Genomic Map