siRNA Details
| Gene Symbol: H3F3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005324.4 | NP_005315.1 | 4 | 1238 | ||
| GenBank mRNA | AF218029.1 | - | 4 | 1062 | ||
| AK054591.1 | - | 1 | 55 | |||
| AK130772.1 | - | 4 | 1081 | |||
| AK304607.1 | - | 4 | 1012 | |||
| BC006497.2 | - | 4 | 1066 | 2959756 | ||
| BC012813.2 | - | 4 | 1066 | 2959756 | ||
| BC017558.1 | - | 4 | 1071 | 3910031 | ||
| BC108701.1 | - | 4 | 1045 | 6144255 | ||
| BX537379.1 | - | 4 | 1082 | |||
| BX647280.1 | - | 4 | 1081 | |||

H3F3B
Genomic Map