siRNA Details
Gene Symbol: CTAG1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001327.2 | NP_001318.1 | 2, 3 | 468 | ||
GenBank mRNA | AF038567.1 | - | 2, 3 | 503 | ||
AJ003149.1 | - | 2, 3 | 465 | |||
AJ275978.1 | - | 2 | 576 | |||
BC130362.1 | - | 2, 3 | 472 | 40146393 | ||
BC130364.1 | - | 2, 3 | 472 | 40146395 | ||
U87459.1 | - | 2, 3 | 468 |
Gene Symbol: CTAG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139250.1 | NP_640343.1 | 2, 3 | 468 |
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020994.4 | NP_066274.2 | 2 | 708 | ||
NM_172377.4 | NP_758965.2 | 2, 3 | 479 | |||
GenBank mRNA | AJ012833.1 | - | 2, 3 | 384 | ||
AJ012834.1 | - | 2, 3 | 468 | |||
AJ012835.1 | - | 2 | 699 | |||
AJ223040.1 | - | 2 | 708 | |||
AJ223041.1 | - | 2, 3 | 467 | |||
AY679088.1 | - | 2, 3 | 467 | |||
AY679089.1 | - | 2, 3 | 467 | |||
BC002833.2 | - | 2 | 662 | 3638129 | ||
BC113998.1 | - | 2, 3 | 459 | 40081511 | ||
BC114933.1 | - | 3, 4 | 242 | |||
BC128045.1 | - | 2, 3 | 459 | 40081514 |
