siRNA Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 4 | 969 | ||
| GenBank mRNA | AK026575.1 | - | 1 | 167 | ||
| AK172732.1 | - | 4 | 834 | |||
| BX537440.1 | - | 4 | 856 | |||
| BX648972.1 | - | 4 | 853 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSBP1| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 4 | 969 | ||
| GenBank mRNA | AK026575.1 | - | 1 | 167 | ||
| AK172732.1 | - | 4 | 834 | |||
| BX537440.1 | - | 4 | 856 | |||
| BX648972.1 | - | 4 | 853 | |||