siRNA Details
| Gene Symbol: KRTAP8-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175857.3 | NP_787053.1 | 1 | 439 | ||
| GenBank mRNA | AJ457064.1 | - | 1 | 439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP8-1| Gene Symbol: KRTAP8-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175857.3 | NP_787053.1 | 1 | 439 | ||
| GenBank mRNA | AJ457064.1 | - | 1 | 439 | ||