siRNA Details
| Gene Symbol: RUFY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037442.3 | NP_001032519.1 | 9 | 1702 | ||
| NM_001130709.1 | NP_001124181.1 | 9 | 1150 | |||
| NM_001291993.1 | NP_001278922.1 | 10 | 1094 | |||
| NM_001291994.1 | NP_001278923.1 | 10 | 1756 | |||
| NM_014961.4 | NP_055776.1 | 9 | 1702 | |||
| XM_005265656.2 | XP_005265713.1 | 10 | 1740 | |||
| XM_005265658.1 | XP_005265715.1 | 9 | 1016 | |||
| XM_011531750.2 | XP_011530052.1 | 10 | 1338 | |||
| XM_011531751.2 | XP_011530053.1 | 9 | 1351 | |||
| XM_011531752.2 | XP_011530054.1 | 9 | 1351 | |||
| XM_011531753.2 | XP_011530055.1 | 9 | 1351 | |||
| XM_011531754.2 | XP_011530056.1 | 9 | 1351 | |||
| XM_011531755.2 | XP_011530057.1 | 9 | 1316 | |||
| XM_011531756.2 | XP_011530058.1 | 9 | 1316 | |||
| XM_017007890.1 | XP_016863379.1 | 9 | 1351 | |||
| XM_017007891.1 | XP_016863380.1 | 10 | 1048 | |||
| XM_017007892.1 | XP_016863381.1 | 9 | 1016 | |||
| GenBank mRNA | AB020678.1 | - | 9 | 1508 | ||
| AF112221.1 | - | 9 | 932 | |||
| AK000911.1 | - | 9 | 956 | |||
| AK294421.1 | - | 9 | 887 | |||
| AK296501.1 | - | 10 | 943 | |||
| AK302637.1 | - | 10 | 970 | |||
| BC051716.1 | - | 9 | 1570 | 5265230 | ||
| BC053837.1 | - | 9 | 1532 | |||
| BC071988.1 | - | 9 | 1505 | |||
| BC098433.1 | - | 9 | 1532 | |||
| DB482497.1 | - | 3 | 168 | |||

RUFY3
Genomic Map