siRNA Details
| Gene Symbol: KRTAP19-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181610.1 | NP_853641.1 | 1 | 137 | ||
| GenBank mRNA | AB096945.1 | - | 1 | 160 | ||
| BC117199.1 | - | 1 | 181 | 40125750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP19-4| Gene Symbol: KRTAP19-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181610.1 | NP_853641.1 | 1 | 137 | ||
| GenBank mRNA | AB096945.1 | - | 1 | 160 | ||
| BC117199.1 | - | 1 | 181 | 40125750 | ||