siRNA Details
| Gene Symbol: ARL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040025.1 | NP_001035114.1 | 4 | 495 | ||
| GenBank mRNA | AK095847.1 | - | 2 | 1623 | ||
| AK304718.1 | - | 4 | 334 | |||
| BC042380.1 | - | 4 | 333 | |||
| BC063687.1 | - | 4 | 297 | |||
| BC105076.1 | - | 4 | 495 | 8144079 | ||
| BC105078.1 | - | 4 | 495 | 8144081 | ||

ARL16
Genomic Map