siRNA Details
| Gene Symbol: TMEM173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301738.1 | NP_001288667.1 | 6 | 1003 | ||
| NM_198282.3 | NP_938023.1 | 8 | 1431 | |||
| XM_005268445.3 | XP_005268502.1 | 7 | 1174 | |||
| XM_011537640.2 | XP_011535942.1 | 7 | 1134 | |||
| GenBank mRNA | AK095896.1 | - | 6 | 2430 | ||
| AK129800.1 | - | 2 | 285 | |||
| AX775913.1 | - | 8 | 1339 | |||
| BC047779.1 | - | 8 | 1348 | 5762441 | ||
| FJ222241.1 | - | 6 | 1097 | |||

TMEM173
Genomic Map