siRNA Details
| Gene Symbol: KLRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198508.2 | NP_940910.1 | 3, 4 | 1082 | ||
| GenBank mRNA | AK126174.1 | - | 3, 4 | 1087 | ||
| BC104963.1 | - | 3, 4 | 1047 | 8143966 | ||
| BC104965.1 | - | 3, 4 | 1047 | 8143968 | ||

KLRG2
Genomic Map