siRNA Details
| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145020.1 | NP_001138492.1 | 2 | 169 | ||
| XM_006715088.3 | XP_006715151.1 | 2 | 338 | |||
| XM_011514575.2 | XP_011512877.1 | 2 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6orf52| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145020.1 | NP_001138492.1 | 2 | 169 | ||
| XM_006715088.3 | XP_006715151.1 | 2 | 338 | |||
| XM_011514575.2 | XP_011512877.1 | 2 | 338 | |||