siRNA Details
| Gene Symbol: SBNO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100122.1 | NP_001093592.1 | 23 | 2811 | ||
| NM_014963.2 | NP_055778.2 | 26 | 3143 | |||
| XM_011527804.2 | XP_011526106.1 | 26 | 3116 | |||
| GenBank mRNA | AB023180.1 | - | 26 | 3120 | ||
| AK074102.1 | - | 22 | 2603 | |||
| AK125139.1 | - | 23 | 2758 | |||
| AK292407.1 | - | 26 | 3143 | |||
| BC106021.1 | - | 3 | 215 | |||

SBNO2
Genomic Map