siRNA Details
Gene Symbol: LCE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276331.1 | NP_001263260.1 | 3 | 443 | ||
NM_178351.3 | NP_848128.1 | 2 | 533 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LCE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276331.1 | NP_001263260.1 | 3 | 443 | ||
NM_178351.3 | NP_848128.1 | 2 | 533 |