siRNA Details
| Gene Symbol: SVBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199342.3 | NP_955374.1 | 3 | 517 | ||
| XM_017001226.1 | XP_016856715.1 | 3 | 520 | |||
| GenBank mRNA | BC029427.1 | - | 3 | 381 | 4809776 | |
| BQ712089.1 | - | 3 | 517 | |||

SVBP
Genomic Map