siRNA Details
| Gene Symbol: FLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014342.2 | NP_001014364.1 | 3 | 6777 | ||
| XM_011509531.2 | XP_011507833.1 | 4 | 6343 | |||
| GenBank mRNA | AY827490.1 | - | 3 | 6778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLG2| Gene Symbol: FLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014342.2 | NP_001014364.1 | 3 | 6777 | ||
| XM_011509531.2 | XP_011507833.1 | 4 | 6343 | |||
| GenBank mRNA | AY827490.1 | - | 3 | 6778 | ||