siRNA Details
| Gene Symbol: CCDC188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243537.1 | NP_001230466.1 | 1, 2 | 177 | ||
| XM_005261238.3 | XP_005261295.1 | 4, 5 | 1622 | |||
| XM_005261240.3 | XP_005261297.1 | 4, 5 | 1621 | |||
| XM_005261241.3 | XP_005261298.1 | 3, 4 | 1514 | |||
| XM_011530170.2 | XP_011528472.1 | 4, 5 | 1622 | |||
| XM_011530171.2 | XP_011528473.1 | 3, 4 | 1517 | |||
| GenBank mRNA | BU579471.1 | - | 1, 2 | 179 | ||

CCDC188
Genomic Map