siRNA Details
| Gene Symbol: LAMTOR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008395.3 | NP_001008396.1 | 4 | 364 | ||
| NM_001318236.1 | NP_001305165.1 | 4 | 381 | |||
| NM_001318237.1 | NP_001305166.1 | 3 | 283 | |||
| NR_134526.1 | 4 | 350 | ||||
| NR_134527.1 | 5 | 406 | ||||
| NR_134528.1 | 4 | 363 | ||||
| NR_134529.1 | 4 | 343 | ||||
| NR_134530.1 | 4 | 380 | ||||
| XM_017012198.1 | XP_016867687.1 | 6 | 751 | |||
| XM_017012199.1 | XP_016867688.1 | 6 | 768 | |||
| GenBank mRNA | AJ714519.1 | - | 4 | 344 | ||
| BC063401.1 | - | 4 | 344 | 5194116 | ||
| BC105609.1 | - | 4 | 354 | 30321610 | ||
| BC130553.1 | - | 4 | 311 | 40146584 | ||
| BC130559.1 | - | 4 | 311 | 40146590 | ||
| BE733059.1 | - | 3 | 273 | |||
| BF978089.1 | - | 362 | ||||
| BG706819.1 | - | 4 | 364 | |||
| BG715626.1 | - | 4 | 359 | |||
| BG819624.1 | - | 4 | 324 | |||
| BI598677.1 | - | 4 | 367 | |||
| BI771662.1 | - | 3 | 269 | |||
| DB498044.1 | - | 4 | 359 | |||
| DB539334.2 | - | 3 | 286 | |||

LAMTOR4
Genomic Map