siRNA Details
| Gene Symbol: FAM221B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_052026.1 | 4 | 1151 | |||
| GenBank mRNA | BX648702.1 | - | 4 | 1151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM221B| Gene Symbol: FAM221B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_052026.1 | 4 | 1151 | |||
| GenBank mRNA | BX648702.1 | - | 4 | 1151 | ||