siRNA Details
| Gene Symbol: WDR47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142550.1 | NP_001136022.1 | 11 | 2482 | ||
| NM_001142551.1 | NP_001136023.1 | 11 | 2458 | |||
| NM_014969.5 | NP_055784.3 | 11 | 2461 | |||
| XM_006710460.3 | XP_006710523.1 | 11 | 2453 | |||
| XM_011541027.2 | XP_011539329.1 | 11 | 2471 | |||
| XM_011541028.2 | XP_011539330.1 | 11 | 2450 | |||
| XM_011541029.1 | XP_011539331.1 | 11 | 2474 | |||
| XM_011541030.1 | XP_011539332.1 | 11 | 2122 | |||
| XM_011541031.1 | XP_011539333.1 | 9 | 1949 | |||
| XM_017000696.1 | XP_016856185.1 | 11 | 2471 | |||
| XM_017000697.1 | XP_016856186.1 | 11 | 2119 | |||
| GenBank mRNA | AB020700.1 | - | 11 | 2305 | ||
| AK074616.1 | - | 4 | 485 | |||
| AK225781.1 | - | 11 | 2242 | |||
| AK289789.1 | - | 11 | 2335 | |||
| AK294998.1 | - | 8 | 1854 | |||
| BC034964.2 | - | 11 | 2346 | 4823902 | ||
| BC039254.1 | - | 10 | 2248 | 5287954 | ||

WDR47
Genomic Map