siRNA Details
| Gene Symbol: OR2A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005328.1 | NP_001005328.1 | 1 | 776 | ||
| GenBank mRNA | BC136704.1 | - | 1 | 870 | 9020694 | |
| BC136711.1 | - | 1 | 870 | 9020701 | ||
| Gene Symbol: OR2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030908.2 | NP_112170.1 | 1 | 776 | ||
| GenBank mRNA | BC120953.2 | - | 1 | 776 | 40115181 | |
| BC120954.1 | - | 1 | 776 | 40115185 | ||
| BI020419.1 | - | 1 | 144 | |||

OR2A7
Genomic Map