siRNA Details
| Gene Symbol: KRTAP5-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004325.1 | NP_001004325.1 | 1 | 215 | ||
| GenBank mRNA | AB126071.1 | - | 1 | 223 | ||
| AJ628245.1 | - | 1 | 215 | |||
| BC132882.1 | - | 1 | 194 | 40146904 | ||
| BC132908.1 | - | 1 | 194 | 40146930 | ||

KRTAP5-2
Genomic Map