siRNA Details
| Gene Symbol: MMRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007351.2 | NP_031377.2 | 2 | 741 | ||
| XM_005262856.2 | XP_005262913.1 | 2 | 766 | |||
| XM_017007893.1 | XP_016863382.1 | 3 | 982 | |||
| GenBank mRNA | AK125557.1 | - | 3 | 688 | ||
| AK302421.1 | - | 2 | 120 | |||
| AK313566.1 | - | 2 | 766 | |||
| BC063848.1 | - | 2 | 741 | 6180674 | ||

MMRN1
Genomic Map