siRNA Details
Gene Symbol: LINC00851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034167.1 | 2 | 156 | |||
GenBank mRNA | BC033985.1 | - | 2 | 161 | 5295734 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034167.1 | 2 | 156 | |||
GenBank mRNA | BC033985.1 | - | 2 | 161 | 5295734 |