siRNA Details
| Gene Symbol: OR2W5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004698.2 | NP_001004698.1 | 1 | 800 | ||
| GenBank mRNA | BC137203.1 | - | 1 | 800 | 9021200 | |
| BC137204.1 | - | 1 | 800 | 9021201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2W5| Gene Symbol: OR2W5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004698.2 | NP_001004698.1 | 1 | 800 | ||
| GenBank mRNA | BC137203.1 | - | 1 | 800 | 9021200 | |
| BC137204.1 | - | 1 | 800 | 9021201 | ||