siRNA Details
| Gene Symbol: ARHGEF35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003702.2 | NP_001003702.2 | 2 | 1701 | ||
| GenBank mRNA | AK125680.1 | - | 2 | 1646 | ||
| BC136868.1 | - | 2 | 1560 | 9020859 | ||
| BC136878.1 | - | 2 | 1560 | 9020869 | ||
| DQ786206.1 | - | 1 | 319 | |||
| EF560743.1 | - | 2 | 1580 | |||

ARHGEF35
Genomic Map