siRNA Details
Gene Symbol: ZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207341.3 | NP_997224.2 | 3 | 679 | ||
GenBank mRNA | EU794684.1 | - | 3 | 679 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207341.3 | NP_997224.2 | 3 | 679 | ||
GenBank mRNA | EU794684.1 | - | 3 | 679 |