siRNA Details
| Gene Symbol: TONSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013432.4 | NP_038460.4 | 21 | 3344 | ||
| XM_011517048.2 | XP_011515350.1 | 14 | 2443 | |||
| XM_011517049.2 | XP_011515351.1 | 14 | 2404 | |||
| GenBank mRNA | AB209126.1 | - | 13 | 4304 | ||
| AK097406.1 | - | 1 | 232 | |||
| BC008782.2 | - | 20 | 3316 | 3628374 | ||

TONSL
Genomic Map