siRNA Details
| Gene Symbol: SPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012508.3 | NP_001012526.2 | 2 | 1109 | ||
| GenBank mRNA | BC040198.1 | - | 2 | 1069 | 4816858 | |
| CD580523.2 | - | 1 | 403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRN| Gene Symbol: SPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012508.3 | NP_001012526.2 | 2 | 1109 | ||
| GenBank mRNA | BC040198.1 | - | 2 | 1069 | 4816858 | |
| CD580523.2 | - | 1 | 403 | |||