siRNA Details
| Gene Symbol: SIDT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040455.1 | NP_001035545.1 | 18 | 1765 | ||
| GenBank mRNA | AF151799.1 | - | 19 | 1870 | ||
| AK075137.1 | - | 9 | 743 | |||
| AK292501.1 | - | 18 | 2245 | |||
| AK292974.1 | - | 18 | 1756 | |||
| AK301501.1 | - | 9 | 961 | |||
| AK303023.1 | - | 18 | 1721 | |||
| AY358442.1 | - | 18 | 1774 | |||
| BC114522.1 | - | 19 | 1785 | 40035836 | ||
| BX648437.1 | - | 10 | 863 | |||
| KC109742.1 | - | 18 | 1721 | |||

SIDT2
Genomic Map