siRNA Details
| Gene Symbol: PCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006198.2 | NP_006189.2 | 3 | 444 | ||
| GenBank mRNA | AK289964.1 | - | 3 | 427 | ||
| AM183336.1 | - | 3 | 382 | |||
| BC013791.1 | - | 3 | 420 | 4345740 | ||
| U52969.1 | - | 3 | 444 | |||
| X93349.1 | - | 3 | 421 | |||

PCP4
Genomic Map