siRNA Details
| Gene Symbol: FAM53C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135647.1 | NP_001129119.1 | 5 | 2147 | ||
| NM_016605.2 | NP_057689.1 | 5 | 2122 | |||
| GenBank mRNA | AF251040.1 | - | 5 | 1858 | ||
| AK027412.1 | - | 3 | 1013 | |||
| BC000259.1 | - | 5 | 1864 | 3357346 | ||

FAM53C
Genomic Map