siRNA Details
| Gene Symbol: CHIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039840.2 | NP_001034929.2 | 5 | 724 | ||
| XM_017029583.1 | XP_016885072.1 | 6 | 405 | |||
| GenBank mRNA | AK297768.1 | - | 5 | 581 | ||
| AL832095.1 | - | 5 | 364 | |||
| BC127728.1 | - | 5 | 635 | 40127525 | ||
| BC136691.1 | - | 5 | 629 | 9020681 | ||
| BC136692.1 | - | 5 | 629 | 9020682 | ||

CHIC1
Genomic Map