siRNA Details
| Gene Symbol: MRPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018997.3 | NP_061870.1 | 2 | 623 | ||
| NM_031901.5 | NP_114107.1 | 3 | 187 | |||
| GenBank mRNA | AB049955.1 | - | 3 | 160 | ||
| AF182417.1 | - | 2 | 132 | |||
| AK058101.1 | - | 2 | 789 | |||
| AK289467.1 | - | 3 | 174 | |||
| BC004566.2 | - | 3 | 156 | 3633098 | ||
| BC006554.2 | - | 2 | 528 | 2819715 | ||
| BI755132.1 | - | 2 | 580 | |||
| HY161691.1 | - | 3 | 188 | |||
| U79258.1 | - | 2 | 544 | |||

MRPS21
Genomic Map