siRNA Details
| Gene Symbol: AKIRIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018064.3 | NP_060534.1 | 3, 4 | 1109 | ||
| GenBank mRNA | AK222701.1 | - | 3, 4 | 1074 | ||
| BC000764.2 | - | 3, 4 | 1050 | 2822886 | ||
| BC003042.1 | - | 3, 4 | 1050 | 2822886 | ||
| BC005051.1 | - | 3, 4 | 787 | 2820292 | ||

AKIRIN2
Genomic Map